4-Chlorobenzoyl-L-tryptophan calcium salt ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C0849-25G C0849-5G
TCIAC0849-25GEA 278 EUR
TCIAC0849-25G TCIAC0849-5G
4-Chlorobenzoyl-L-tryptophan calcium salt ≥98.0% (by HPLC)
4-Chlorobenzoyl-L-tryptophan calcium salt
Formule: C₁₈H₁₅ClN₂O₃
MDL Nummer: MFCD00036700
CAS nummer: 56116-62-2
EINECS: 260-001-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 97.0 % (after drying)
Solubility in Methanol almost transparency
Drying loss max. 3.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR