Vinyl crotonate ≥99.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Crotonic Acid Vinyl Ester

Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

C0897-25ML C0897-500ML
TCIAC0897-25MLEA 19.8 EUR
TCIAC0897-25ML TCIAC0897-500ML
Vinyl crotonate ≥99.0% (by GC) stabilised
Vinyl crotonate
Formule: C₆H₈O₂
Kookpunt: 134°C
Vlampunt: 32°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009288
CAS nummer: 14861-06-4
EINECS: 238-930-6
UN: 3272
ADR: 3,III
Merck Index: 14,02597

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Acid Value max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR