2-Methoxyethyl chloroformate ≥93.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0928-25ML C0928-5ML
TCIAC0928-25MLEA 143 EUR
TCIAC0928-25ML TCIAC0928-5ML
2-Methoxyethyl chloroformate ≥93.0% (by GC, titration analysis)
2-Methoxyethyl chloroformate
Formule: C₄H₇ClO₃
MW: 138.55 g/mol
Kookpunt: 59°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00058932
CAS nummer: 628-12-6
EINECS: 211-026-9
UN: 2742
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b3e1e808da5451c75f5d09730bfb3a26087793&vid=34b3e1e808da5451c75f5d09730bfb3a26087793&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %
Purity (Argentmetric Titration) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR