Chromeazurol S (Mordant blue 29)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,6-Dichlorodimethyl-sulphoxy-fuchsonedicarboxylic acid sodium salt, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid,trisodium salt, Mordant Blue 29, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid, trisodium salt, Chrome Azurol S

C0979-25G C0979-5G
TCIAC0979-25GEA 130 EUR
TCIAC0979-25G TCIAC0979-5G
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)
Formule: C₂₃H₁₆Cl₂O₉S
MDL Nummer: MFCD00001615
CAS nummer: 1667-99-8
EINECS: 216-787-0

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 425.0 to 435.0 nm (pH7.0 phosphate buffer)
Absorbance min. 0.500 (0.02g/L,pH7.0 phosphate buffer,425.0 to 435.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %
Ignition residue (Sulfate) 35.0 to 45.0% (calcd.on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR