N-Chloroacetylglycine ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Chloroac-Gly-OH, N-Chloroacetylglycine, 2-(2-Chloroacetamido)acetic acid

C0980-1G C0980-25G
TCIAC0980-1GEA 37.4 EUR
TCIAC0980-1G TCIAC0980-25G
N-Chloroacetylglycine ≥99.0% (by titrimetric analysis)
N-Chloroacetylglycine
Formule: C₄H₆ClNO₃
MDL Nummer: MFCD00021733
CAS nummer: 6319-96-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 103.0 to 107.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR