Ethyl-2-cyanopropionate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethyl 2-cyanopropanoate, 2-Cyanopropionic Acid Ethyl Ester
C1083-25ML
C1083-5ML
TCIAC1083-25MLEA
137
EUR
TCIAC1083-25ML
TCIAC1083-5ML
Ethyl-2-cyanopropionate ≥97.0% (by GC)
Ethyl-2-cyanopropionate
Formule:
C₆H₉NO₂ MW: 127.14 g/mol Kookpunt: 193°C Vlampunt: 96°C |
MDL Nummer:
MFCD00013812 CAS nummer: 1572-99-2 EINECS: 216-393-9 UN: 3276 ADR: 6.1 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...