3-Chloropropyldimethoxymethylsilane ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C1168-25G
C1168-5G
TCIAC1168-25GEA
109
EUR
TCIAC1168-25G
TCIAC1168-5G
3-Chloropropyldimethoxymethylsilane ≥95.0% (by GC)
3-Chloropropyldimethoxymethylsilane
Formule:
C₆H₁₅ClO₂Si Kookpunt: 185°C Vlampunt: 49°C |
MDL Nummer:
MFCD00053912 CAS nummer: 18171-19-2 EINECS: 242-056-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...