(3-Cyanopropyl)dimethylchlorosilane ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Chloro(3-cyanopropyl)dimethylsilane
C1207-25ML C1207-5ML
TCIAC1207-25MLEA 344 EUR
TCIAC1207-25ML TCIAC1207-5ML
(3-Cyanopropyl)dimethylchlorosilane ≥95.0% (by GC)
(3-Cyanopropyl)dimethylchlorosilane
Formule: C₆H₁₂ClNSi
Kookpunt: 109°C
Vlampunt: 108°C
MDL Nummer: MFCD00001969
CAS nummer: 18156-15-5
EINECS: 242-039-8
UN: 2987
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %