2-(4-Chlorobutoxy)tetrahydropyran ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C1279-25G C1279-5G C1279-1G
TCIAC1279-25GEA 300 EUR
TCIAC1279-25G TCIAC1279-5G TCIAC1279-1G
2-(4-Chlorobutoxy)tetrahydropyran ≥95.0% (by GC)
2-(4-Chlorobutoxy)tetrahydropyran
Formule: C₉H₁₇ClO₂
Kookpunt: 131°C
MDL Nummer: MFCD00142812
CAS nummer: 41302-05-0
EINECS: 255-305-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30d6a2516c5c3de4fb9660520508bd72154afe8&vid=b30d6a2516c5c3de4fb9660520508bd72154afe8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR