Ethyl-2-cyano-2-methylpropionate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1546-25G C1546-5G
TCIAC1546-25GEA 145 EUR
TCIAC1546-25G TCIAC1546-5G
Ethyl-2-cyano-2-methylpropionate ≥98.0% (by GC)
Ethyl-2-cyano-2-methylpropionate
Formule: C₇H₁₁NO₂
Kookpunt: 185°C
MDL Nummer: MFCD00034667
CAS nummer: 1572-98-1
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %