Chloromethyl polystyrene resin 1,6 - 1,8 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C1643-25G C1643-5G
TCIAC1643-25GEA 194 EUR
TCIAC1643-25G TCIAC1643-5G
Chloromethyl polystyrene resin 1,6 - 1,8 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Chloromethyl polystyrene resin
MDL Nummer: MFCD00146406
CAS nummer: 55844-94-5
Order Now
Specification Test Results
Loading 1.5 to 1.8 mmol/g