1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥94.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1857-1G C1857-5G
TCIAC1857-1GEA 46.2 EUR
TCIAC1857-1G TCIAC1857-5G
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥94.0% (by GC)
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Formule: C₁₀H₁₀ClF₁₃Si
Kookpunt: 80°C
Vlampunt: 52°C
MDL Nummer: MFCD00042359
CAS nummer: 102488-47-1
UN: 2986
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %