3-(Carbobenzoxyamino)-1-propanol ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Benzyl N-(3-Hydroxypropyl)carbamate, N-(3-Hydroxypropyl)carbamic Acid Benzyl Ester, 3-(Cbz-amino)-1-propanol, Benzyl (3-hydroxypropyl)carbamate

C1932-25G C1932-5G
TCIAC1932-25GEA 143 EUR
TCIAC1932-25G TCIAC1932-5G
3-(Carbobenzoxyamino)-1-propanol ≥98.0% (by HPLC, total nitrogen)
3-(Carbobenzoxyamino)-1-propanol
Formule: C₁₁H₁₅NO₃
MDL Nummer: MFCD01321351
CAS nummer: 34637-22-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR