Climbazole ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone

C2025-25G C2025-500G
TCIAC2025-25GEA 40.7 EUR
TCIAC2025-25G TCIAC2025-500G
Climbazole ≥98.0% (by HPLC, titration analysis)
Climbazole
Formule: C₁₅H₁₇ClN₂O₂
CAS nummer: 38083-17-9
EINECS: 253-775-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR