Cloxacillin sodium salt monohydrate ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C2244-1G
C2244-5G
TCIAC2244-1GEA
42.9
EUR
TCIAC2244-1G
TCIAC2244-5G
Cloxacillin sodium salt monohydrate ≥98.0% (by HPLC)
Cloxacillin sodium salt monohydrate
Formule:
C₁₉H₁₈ClN₃O₅S Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00150735 CAS nummer: 7081-44-9 |
Specification Test Results
Appearance | White to Orange to Green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +160 to +169 deg (C=1, H2O) (calcd.on anh.substance) |
Water | 3.0 to 5.3 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...