cis-1,3-Cyclohexanediol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C2322-1G C2322-5G
TCIAC2322-1GEA 645 EUR
TCIAC2322-1G TCIAC2322-5G
cis-1,3-Cyclohexanediol ≥98.0% (by GC)
cis-1,3-Cyclohexanediol
Formule: C₆H₁₂O₂
Kookpunt: 125°C
CAS nummer: 823-18-7
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C