Benzyl-3-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-Benzyloxycarbonyl-3-pyrrolidone, 1-Carbobenzoxy-3-pyrrolidone, 1-Cbz-3-pyrrolidinone, 1-Cbz-3-pyrrolidone, Benzyl 3-oxopyrrolidine-1-carboxylate, 1-Benzyloxycarbonyl-3-pyrrolidinone, 1-Carbobenzoxy-3-pyrrolidinone
C2437-1G
C2437-5G
TCIAC2437-1GEA
37.4
EUR
TCIAC2437-1G
TCIAC2437-5G
Benzyl-3-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
Benzyl-3-oxopyrrolidine-1-carboxylate
Formule:
C₁₂H₁₃NO₃ MW: 219.2365 g/mol |
MDL Nummer:
MFCD03001711 CAS nummer: 130312-02-6 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...