2-Chloro-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C2467-1G C2467-5G
TCIAC2467-1GEA 42.9 EUR
TCIAC2467-1G TCIAC2467-5G
2-Chloro-4-(trifluoromethyl)pyrimidine ≥98.0% (by GC)
2-Chloro-4-(trifluoromethyl)pyrimidine
Formule: C₅H₂ClF₃N₂
Kookpunt: 60°C
Vlampunt: 49°C
MDL Nummer: MFCD00115111
CAS nummer: 33034-67-2
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %