Cefsulodin sodium salt hydrate ≥95.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C2598-1G C2598-200MG
TCIAC2598-1GEA 269 EUR
TCIAC2598-1G TCIAC2598-200MG
Cefsulodin sodium salt hydrate ≥95.0% (by HPLC, total nitrogen)
Cefsulodin sodium salt hydrate
Formule: C₂₂H₁₉N₄NaO₈S₂·xH₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD07793338
CAS nummer: 1426397-23-0
EINECS: 257-692-4
Merck Index: 14,01945

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +16.0 to +20.0 deg (C=1, H2O) (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR