Chloromethyl isopropyl carbonate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2626-25G
C2626-100G
TCIAC2626-25GEA
60.5
EUR
TCIAC2626-25G
TCIAC2626-100G
Chloromethyl isopropyl carbonate ≥98.0% (by GC)
Chloromethyl isopropyl carbonate
Formule:
C₅H₉ClO₃ |
MDL Nummer:
MFCD07375443 CAS nummer: 35180-01-9 UN: 2924 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...