3-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3-Chlorobenzoic hydrazide
C2648-5G
C2648-25G
TCIAC2648-5GEA
38.5
EUR
TCIAC2648-5G
TCIAC2648-25G
3-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
3-Chlorobenzhydrazide
Formule:
C₇H₇ClN₂O MW: 170.6 g/mol |
MDL Nummer:
MFCD00014757 CAS nummer: 1673-47-8 EINECS: 216-812-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...