Cyclopenta[fg]tetracene-1,2-dione
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C2669-200MG
C2669-1G
TCIAC2669-200MGEA
120
EUR
TCIAC2669-200MG
TCIAC2669-1G
Cyclopenta[fg]tetracene-1,2-dione
Cyclopenta[fg]tetracene-1,2-dione
Formule:
C₂₀H₁₀O₂ |
CAS nummer:
1334510-66-5 |
Specification Test Results
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Elemental analysis (Carbon) | 83.00 to 87.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...