5-Methylpicolinonitrile ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2706-1G
C2706-5G
TCIAC2706-1GEA
31.9
EUR
TCIAC2706-1G
TCIAC2706-5G
5-Methylpicolinonitrile ≥98.0% (by GC, titration analysis)
5-Methylpicolinonitrile
Formule:
C7H6N2 Kookpunt: 140°C |
MDL Nummer:
MFCD06200830 CAS nummer: 1620-77-5 EINECS: 216-589-4 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...