Chloro(dimethylsulphide)gold(I) ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (Dimethylsulphide)gold(I) Chloride

C2719-200MG C2719-1G
TCIAC2719-200MGEA 64.9 EUR
TCIAC2719-200MG TCIAC2719-1G
Chloro(dimethylsulphide)gold(I) ≥97.0% (by titrimetric analysis)
Chloro(dimethylsulphide)gold(I)
Formule: C₂H₆AuClS
MW: 294.55 g/mol
Smeltpunt: 160°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00192090
CAS nummer: 29892-37-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR