Cefoperazone ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C2768-5G C2768-25G
TCIAC2768-5GEA 84.7 EUR
TCIAC2768-5G TCIAC2768-25G
Cefoperazone ≥98.0% (by HPLC, titration analysis)
Cefoperazone
Formule: C₂₅H₂₇N₉O₈S₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00865067
CAS nummer: 62893-19-0
EINECS: 263-749-4
Merck Index: 14,01930

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR