Cephalothin ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2769-5G
C2769-25G
TCIAC2769-5GEA
35.2
EUR
TCIAC2769-5G
TCIAC2769-25G
Cephalothin ≥98.0% (by HPLC, titration analysis)
Cephalothin
Formule:
C₁₆H₁₆N₂O₆S₂ |
MDL Nummer:
MFCD00242614 CAS nummer: 153-61-7 EINECS: 205-815-7 Merck Index: 14,01982 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...