2-Chloro-6-methylpyrazine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C2777-1G C2777-5G
TCIAC2777-1GEA 83.6 EUR
TCIAC2777-1G TCIAC2777-5G
2-Chloro-6-methylpyrazine ≥98.0% (by GC)
2-Chloro-6-methylpyrazine
Formule: C₅H₅ClN₂
MW: 128.5596 g/mol
Kookpunt: 83°C
MDL Nummer: MFCD00055032
CAS nummer: 38557-71-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR