Cesium green ≥85.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C2806-50MG
C2806-250MG
TCIAC2806-50MGEA
127
EUR
TCIAC2806-50MG
TCIAC2806-250MG
Cesium green ≥85.0% (by HPLC)
Cesium green
Formule:
C₃₂H₃₃NO₈ |
CAS nummer:
1611477-45-2 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...