Chitosan oligosaccharides
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C2849-25G C2849-100G
TCIAC2849-25GEA 59.4 EUR
TCIAC2849-25G TCIAC2849-100G
Chitosan oligosaccharides
Chitosan oligosaccharides
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Total Nitrogen 6.0 to 7.0 %
Water max. 10.0 %