4-Chloro-1-iodo-2-nitrobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C2932-1G C2932-5G
TCIAC2932-1GEA 58.3 EUR
TCIAC2932-1G TCIAC2932-5G
4-Chloro-1-iodo-2-nitrobenzene ≥98.0% (by GC)
4-Chloro-1-iodo-2-nitrobenzene
Formule: C₆H₃ClINO₂
MDL Nummer: MFCD11226294
CAS nummer: 5446-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28de99670cb83aa05ad135584c1d52f49d0b7240&vid=28de99670cb83aa05ad135584c1d52f49d0b7240&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C