3-Cyclopentylpropionic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C2934-5G
C2934-25G
TCIAC2934-5GEA
24.2
EUR
TCIAC2934-5G
TCIAC2934-25G
3-Cyclopentylpropionic acid ≥98.0% (by GC, titration analysis)
3-Cyclopentylpropionic acid
Formule:
C₈H₁₄O₂ Kookpunt: 123°C Vlampunt: 47°C |
MDL Nummer:
MFCD00001392 CAS nummer: 140-77-2 EINECS: 205-433-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...