3-Chloro-5-(trifluoromethyl)pyridine ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2951-1G
C2951-5G
TCIAC2951-1GEA
22
EUR
TCIAC2951-1G
TCIAC2951-5G
3-Chloro-5-(trifluoromethyl)pyridine ≥97.0% (by GC)
3-Chloro-5-(trifluoromethyl)pyridine
Formule:
C6H3ClF3N Vlampunt: 54°C |
MDL Nummer:
MFCD00042227 CAS nummer: 85148-26-1 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...