3-Chloro-4-hydroxyphenylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C2969-1G C2969-5G
TCIAC2969-1GEA 47.3 EUR
TCIAC2969-1G TCIAC2969-5G
3-Chloro-4-hydroxyphenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-hydroxyphenylboronic acid
Formule: C₆H₆BClO₃
MDL Nummer: MFCD09258739
CAS nummer: 182344-13-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR