Cerium triacetate hydrate ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate
C2974-25G C2974-500G
TCIAC2974-25GEA 27.5 EUR
TCIAC2974-25G TCIAC2974-500G
Cerium triacetate hydrate ≥98.0% (by titrimetric analysis)
Cerium triacetate hydrate
Formule: C₆H₉CeO₆·H₂O
MW: 335.266 g/mol
Smeltpunt: 308°C
MDL Nummer: MFCD00150533
CAS nummer: 206996-60-3
EINECS: 208-654-0
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 4.0 to 10.0 %