7-Chloro-1-tetralone ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C2976-1G C2976-5G
TCIAC2976-1GEA 45.1 EUR
TCIAC2976-1G TCIAC2976-5G
7-Chloro-1-tetralone ≥95.0% (by GC)
7-Chloro-1-tetralone
Formule: C₁₀H₉ClO
Kookpunt: 115°C
MDL Nummer: MFCD01928386
CAS nummer: 26673-32-5
Order Now
Specification Test Results
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 93.0 to 97.0 deg-C