(3-Carbamoylphenyl)boronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C2992-1G C2992-5G
TCIAC2992-1GEA 79.2 EUR
TCIAC2992-1G TCIAC2992-5G
(3-Carbamoylphenyl)boronic acid (contains varying amounts of Anhydride)
(3-Carbamoylphenyl)boronic acid
Formule: C₇H₈BNO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03411948
CAS nummer: 351422-73-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR