L(-)-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride

C3058-5G C3058-25G
TCIAC3058-5GEA 36.3 EUR
TCIAC3058-5G TCIAC3058-25G
L(-)-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)
L(-)-Carnitine hydrochloride
Formule: C₇H₁₆ClNO₃
MW: 197.66 g/mol
MDL Nummer: MFCD00066100
CAS nummer: 6645-46-1
EINECS: 229-663-6
Merck Index: 14,01849

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C
Specific rotation [a]20/D -21.0 to -25.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR