Coumarin 510 ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,3,6,7-Tetrahydro-10-(3-pyridyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one

C3072-200MG C3072-1G
TCIAC3072-200MGEA 72.6 EUR
TCIAC3072-200MG TCIAC3072-1G
Coumarin 510 ≥98.0% (by HPLC, total nitrogen)
Coumarin 510
Formule: C₂₀H₁₈N₂O₂
MDL Nummer: MFCD00467042
CAS nummer: 87349-92-6

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Molar extinction coefficient min. 34000 (THF, 412 to 417nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR