5-Chloro-4-fluoro-2-nitroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-Amino-3-chlor-4-fluor-6-nitrobenzene

C3141-1G C3141-5G
TCIAC3141-1GEA 30.8 EUR
TCIAC3141-1G TCIAC3141-5G
5-Chloro-4-fluoro-2-nitroaniline ≥98.0% (by GC)
5-Chloro-4-fluoro-2-nitroaniline
Formule: C₆H₄ClFN₂O₂
MW: 190.56 g/mol
MDL Nummer: MFCD00052698
CAS nummer: 104222-34-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR