2-Chloro-6-methylbenzothiazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Chloro-6-methyl-1,3-benzothiazole, 2-Chloro-6-methylbenzo[d]thiazole

C3160-1G C3160-5G
TCIAC3160-1GEA 60.5 EUR
TCIAC3160-1G TCIAC3160-5G
2-Chloro-6-methylbenzothiazole ≥98.0% (by GC)
2-Chloro-6-methylbenzothiazole
Formule: C₈H₆ClNS
MW: 183.66 g/mol
Kookpunt: 152°C
MDL Nummer: MFCD04448821
CAS nummer: 3507-26-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR