Cyclohexene-1-carbonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C3191-1G
C3191-5G
TCIAC3191-1GEA
45.1
EUR
TCIAC3191-1G
TCIAC3191-5G
Cyclohexene-1-carbonitrile ≥98.0% (by GC)
Cyclohexene-1-carbonitrile
Formule:
C₇H₉N Kookpunt: 74°C Vlampunt: 83°C |
MDL Nummer:
MFCD00013772 CAS nummer: 1855-63-6 EINECS: 217-454-2 UN: 3276 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...