3-Chloro-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C3192-5G C3192-25G
TCIAC3192-5GEA 62.7 EUR
TCIAC3192-5G TCIAC3192-25G
3-Chloro-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)
3-Chloro-N,N-dimethylaniline
Formule: C₈H₁₀ClN
Kookpunt: 234°C
MDL Nummer: MFCD00018456
CAS nummer: 6848-13-1
EINECS: 229-935-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR