2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C3208-5G C3208-25G
TCIAC3208-5GEA 94.6 EUR
TCIAC3208-5G TCIAC3208-25G
2-Chloro-5-(trifluoromethyl)benzyl alcohol ≥98.0% (by GC)
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Formule: C₈H₆ClF₃O
MDL Nummer: MFCD01631469
CAS nummer: 64372-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d33dd50bec0d8f7c5bbac49bbef245add8617d6&vid=6d33dd50bec0d8f7c5bbac49bbef245add8617d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR