4-Chloro-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C3212-1G
C3212-5G
TCIAC3212-1GEA
82.5
EUR
TCIAC3212-1G
TCIAC3212-5G
4-Chloro-3-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-nitrophenylboronic acid
Formule:
C₆H₅BClNO₄ |
MDL Nummer:
MFCD02258950 CAS nummer: 151169-67-4 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...