4-Chloro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C3248-200MG
C3248-1G
TCIAC3248-200MGEA
34.1
EUR
TCIAC3248-200MG
TCIAC3248-1G
4-Chloro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-methoxyphenylboronic acid
Formule:
C₇H₈BClO₃ |
MDL Nummer:
MFCD02684317 CAS nummer: 762287-57-0 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...