Carboxymethyldextran sodium salt, M.W. ~ 10000

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C3250-1G C3250-5G
TCIAC3250-1GEA 36.3 EUR
TCIAC3250-1G TCIAC3250-5G
Carboxymethyldextran sodium salt, M.W. ~ 10000
Carboxymethyldextran sodium salt
MDL Nummer: MFCD00146477
CAS nummer: 39422-83-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Content (Sodium) 6.5 to 9.5 % (calcd.on dried substance)
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR