5-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C3286-1G C3286-5G
TCIAC3286-1GEA 30.8 EUR
TCIAC3286-1G TCIAC3286-5G
5-Fluoropyridine-2-carbonitrile ≥98.0% (by GC)
5-Fluoropyridine-2-carbonitrile
Formule: C₆H₃FN₂
MDL Nummer: MFCD08741371
CAS nummer: 327056-62-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR