3'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C3288-1G C3288-5G
TCIAC3288-1GEA 30.8 EUR
TCIAC3288-1G TCIAC3288-5G
3'-Chloro-5'-fluoroacetophenone ≥98.0% (by GC)
3'-Chloro-5'-fluoroacetophenone
Formule: C₈H₆ClFO
MDL Nummer: MFCD04115834
CAS nummer: 842140-52-7
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C