Cyclohexyldiphenylphosphine oxide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C3310-1G C3310-5G
TCIAC3310-1GEA 22 EUR
TCIAC3310-1G TCIAC3310-5G
Cyclohexyldiphenylphosphine oxide ≥98.0% (by GC)
Cyclohexyldiphenylphosphine oxide
Formule: C₁₈H₂₁OP
MDL Nummer: MFCD00014301
CAS nummer: 13689-20-8
EINECS: 237-204-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4e54621317d8dfd25bafeb859c48be14af74acc&vid=b4e54621317d8dfd25bafeb859c48be14af74acc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR