Chloro(hexyl)dimethylsilane ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C3415-5ML
C3415-25ML
TCIAC3415-5MLEA
33
EUR
TCIAC3415-5ML
TCIAC3415-25ML
Chloro(hexyl)dimethylsilane ≥96.0% (by GC)
Chloro(hexyl)dimethylsilane
Formule:
C₈H₁₉ClSi Kookpunt: 184°C |
MDL Nummer:
MFCD01073817 CAS nummer: 3634-59-1 UN: 2987 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...